Learn More
4-Amino-2-chloropyridine, 98%, Thermo Scientific Chemicals
$115.16 - $337.44
Chemical Identifiers
CAS | 14432-12-3 |
---|---|
Molecular Formula | C5H5ClN2 |
Molecular Weight (g/mol) | 128.559 |
MDL Number | MFCD00060089 |
InChI Key | BLBDTBCGPHPIJK-UHFFFAOYSA-N |
Synonym | 4-amino-2-chloropyridine, 2-chloro-4-aminopyridine, 2-chloro-pyridin-4-ylamine, 4-pyridinamine, 2-chloro, 2-chloro-4-pyridinamine, 2-chlorpyridin-4-amin, 4-amino-6-chloropyridine, 2-chloro-4-pyridylamine, 2-chloro-4-amino pyridine, 4-amino-2-chloro pyridine |
PubChem CID | 84432 |
IUPAC Name | 2-chloropyridin-4-amine |
SMILES | C1=CN=C(C=C1N)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2209806
|
Thermo Scientific Chemicals
B2209806 |
5 g |
N/A
|
|
|||||
AAB2209814
|
Thermo Scientific Chemicals
B2209814 |
25 g |
N/A
|
|
|||||
Description
2-Chloropyridin-4-amine (14432-12-3) is used as a medicine, pigment intermediates. It is an additive of plant growth regulator.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14432-12-3 | |
128.559 | |
BLBDTBCGPHPIJK-UHFFFAOYSA-N | |
84432 | |
C1=CN=C(C=C1N)Cl |
C5H5ClN2 | |
MFCD00060089 | |
4-amino-2-chloropyridine, 2-chloro-4-aminopyridine, 2-chloro-pyridin-4-ylamine, 4-pyridinamine, 2-chloro, 2-chloro-4-pyridinamine, 2-chlorpyridin-4-amin, 4-amino-6-chloropyridine, 2-chloro-4-pyridylamine, 2-chloro-4-amino pyridine, 4-amino-2-chloro pyridine | |
2-chloropyridin-4-amine |
Specifications
14432-12-3 | |
153°C (5 mmHg) | |
C5H5ClN2 | |
5 g | |
4-amino-2-chloropyridine, 2-chloro-4-aminopyridine, 2-chloro-pyridin-4-ylamine, 4-pyridinamine, 2-chloro, 2-chloro-4-pyridinamine, 2-chlorpyridin-4-amin, 4-amino-6-chloropyridine, 2-chloro-4-pyridylamine, 2-chloro-4-amino pyridine, 4-amino-2-chloro pyridine | |
BLBDTBCGPHPIJK-UHFFFAOYSA-N | |
2-chloropyridin-4-amine | |
84432 | |
98% | |
4-Amino-2-chloropyridine |
91°C to 94°C | |
Plastic bottle | |
MFCD00060089 | |
108671 | |
Slightly soluble in water. | |
C1=CN=C(C=C1N)Cl | |
128.559 | |
128.56 | |
Crystalline Powder |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
H301+H311+H331-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 238-403-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only