missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Amino-2-(chloromethyl)pyrimidine, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H66760MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Amino-2-(chloromethyl)pyrimidine | |
Colorless to Yellow | |
264°C | |
MFCD10697353 | |
2-chloromethyl pyrimidin-4-amine, 2-chloromethyl-4-amino-pyrimidine, 4-pyrimidinamine, 2-chloromethyl, 2-chloromethyl-4-aminopyrimidine, 4-pyrimidinamine, 2-chloromethyl-9ci, 2-chloromethyl-pyrimidin-4-ylamine, 4-amino-2-chloromethylpyrimidine, 2-chloromethyl 4 amino pyrimidine, 2-chloromethyl-4-pyrimidinamine, 4-pyrimidinamine,2-chloromethyl-9ci | |
C1=CN=C(N=C1N)CCl | |
143.574 | |
143.57 | |
Liquid |
79651-35-7 | |
1.35 g/mL | |
C5H6ClN3 | |
250 mg | |
JWAQENUKTAOJST-UHFFFAOYSA-N | |
2-(chloromethyl)pyrimidin-4-amine | |
21298827 | |
96% |
Chemical Identifiers
79651-35-7 | |
143.574 | |
JWAQENUKTAOJST-UHFFFAOYSA-N | |
21298827 | |
C1=CN=C(N=C1N)CCl |
C5H6ClN3 | |
MFCD10697353 | |
2-chloromethyl pyrimidin-4-amine, 2-chloromethyl-4-amino-pyrimidine, 4-pyrimidinamine, 2-chloromethyl, 2-chloromethyl-4-aminopyrimidine, 4-pyrimidinamine, 2-chloromethyl-9ci, 2-chloromethyl-pyrimidin-4-ylamine, 4-amino-2-chloromethylpyrimidine, 2-chloromethyl 4 amino pyrimidine, 2-chloromethyl-4-pyrimidinamine, 4-pyrimidinamine,2-chloromethyl-9ci | |
2-(chloromethyl)pyrimidin-4-amine |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only