missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-2-chlorobenzoic acid, 98%

Manufacturer:  Alfa Aesar™ A1011606

 View more versions of this product

Catalog No. AAA1011606



Specifications

Specifications

4-Amino-2-chlorobenzoic acid
5g
C7H6ClNO2
2803668
MBDUKNCPOPMRJQ-UHFFFAOYSA-N
4-amino-2-chlorobenzoic acid
17154
171.58
~220°C (decomposition)
2457-76-3
MFCD00007772
2-chloro-4-aminobenzoic acid, benzoic acid, 4-amino-2-chloro, o-chloro-p-aminobenzoic acid, 2-chloro-p-aminobenzoic acid, usaf nb-1, 4-amino-2-chloro-benzoic acid, 4-amino-2-chlorobenzoicacid, unii-y3s6924ia6, wln: zr cg dvq, pubchem3109
C1=CC(=C(C=C1N)Cl)C(=O)O
171.58
CHEBI:59472
98%

Chemical Identifiers

2457-76-3
171.58
MBDUKNCPOPMRJQ-UHFFFAOYSA-N
17154
4-amino-2-chlorobenzoic acid
C7H6ClNO2
MFCD00007772
2-chloro-4-aminobenzoic acid, benzoic acid, 4-amino-2-chloro, o-chloro-p-aminobenzoic acid, 2-chloro-p-aminobenzoic acid, usaf nb-1, 4-amino-2-chloro-benzoic acid, 4-amino-2-chlorobenzoicacid, unii-y3s6924ia6, wln: zr cg dvq, pubchem3109
CHEBI:59472
C1=CC(=C(C=C1N)Cl)C(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DG1575000

TSCA : TSCA

SDS Documents