Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-2-chlorobenzoic acid, 98%

$88.15 - $1,167.74

Chemical Identifiers

Chemical Name or Material 4-Amino-2-chlorobenzoic acid
CAS 2457-76-3
Molecular Formula C7H6ClNO2
Formula Weight 171.58
MDL Number MFCD00007772
Beilstein 2803668
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1011606

  SDS
alfa aesar™
A1011606
5g Each for $88.15

AAA1011614

  SDS
alfa aesar™
A1011614
25g Each for $355.59

AAA1011622

  SDS
alfa aesar™
A1011622
100g Each for $1,167.74
Description & Specifications

Specifications

Formula Weight 171.58
TSCA TSCA
Melting Point ≈220°C (dec.)
CAS 2457-76-3
Beilstein 2803668
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00007772
RTECS Number DG1575000
Quantity 5g
Chemical Name or Material 4-Amino-2-chlorobenzoic acid
Molecular Formula C7H6ClNO2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.