Learn More
4-Amino-2-chlorobenzoic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1011614
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Amino-2-chlorobenzoic acid | |
∼220°C (decomposition) | |
MFCD00007772 | |
2803668 | |
MBDUKNCPOPMRJQ-UHFFFAOYSA-N | |
4-amino-2-chlorobenzoic acid | |
17154 | |
171.58 |
2457-76-3 | |
C7H6ClNO2 | |
25 g | |
2-chloro-4-aminobenzoic acid, benzoic acid, 4-amino-2-chloro, o-chloro-p-aminobenzoic acid, 2-chloro-p-aminobenzoic acid, usaf nb-1, 4-amino-2-chloro-benzoic acid, 4-amino-2-chlorobenzoicacid, unii-y3s6924ia6, wln: zr cg dvq, pubchem3109 | |
C1=CC(=C(C=C1N)Cl)C(=O)O | |
171.58 | |
CHEBI:59472 | |
98% |
Chemical Identifiers
2457-76-3 | |
171.58 | |
MBDUKNCPOPMRJQ-UHFFFAOYSA-N | |
17154 | |
4-amino-2-chlorobenzoic acid |
C7H6ClNO2 | |
MFCD00007772 | |
2-chloro-4-aminobenzoic acid, benzoic acid, 4-amino-2-chloro, o-chloro-p-aminobenzoic acid, 2-chloro-p-aminobenzoic acid, usaf nb-1, 4-amino-2-chloro-benzoic acid, 4-amino-2-chlorobenzoicacid, unii-y3s6924ia6, wln: zr cg dvq, pubchem3109 | |
CHEBI:59472 | |
C1=CC(=C(C=C1N)Cl)C(=O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 219-540-5
RTECSNumber : DG1575000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only