Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-2-chloro-6,7-dimethoxyquinazoline, 98+%

$115.45 - $441.77

Chemical Identifiers

Chemical Name or Material 4-Amino-2-chloro-6,7-dimethoxyquinazoline
CAS 23680-84-4
Molecular Formula C10H10ClN3O2
Formula Weight 239.66
MDL Number MFCD00051734
Beilstein 747463
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1495206

 
alfa aesar™
A1495206
5g Each for $115.45

AAA1495214

 
alfa aesar™
A1495214
25g Each for $441.77
Description & Specifications

Specifications

Formula Weight 239.66
Chemical Name or Material 4-Amino-2-chloro-6,7-dimethoxyquinazoline
Melting Point ≈280°C (dec.)
Quantity 5g
Assay ≥98%
Molecular Formula C10H10ClN3O2
MDL Number MFCD00051734
CAS 23680-84-4
Beilstein 747463
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.