Please login to your online account to display your discounted pricing

4-Amino-2-chloro-6,7-dimethoxyquinazoline, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Amino-2-chloro-6, 7-dimethoxyquinazoline
CAS 23680-84-4
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10H10ClN3O2
Formula Weight 239.66
MDL Number MFCD00051734
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC342540250

  SDS
thermo fisher scientific
342540250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Fluffy Crystalline Powder
Formula Weight 239.66
Quantity 25g
Color Off-white
Melting Point 273° to 278°C
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
CAS 23680-84-4
Chemical Name or Material 4-Amino-2-chloro-6, 7-dimethoxyquinazoline
Molecular Formula C10H10ClN3O2
Assay Percent Range 97%
MDL Number MFCD00051734
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.