Please login to your online account to display your discounted pricing

4-Amino-2-chloro-5-fluoropyrimidine, 98% , ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Amino-2-chloro-5-fluoropyrimidine
CAS 155-10-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H3ClFN3
Formula Weight 147.54
MDL Number MFCD00057344
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC399980010

  SDS
thermo fisher scientific
AC399980010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC399980050

  SDS
thermo fisher scientific
AC399980050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC399982500

  SDS
thermo fisher scientific
AC399982500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 147.54
Assay ≥97.5%
Physical Form Crystalline Powder
Quantity 1g
Color White to light yellow
Melting Point 192° to 196°C
CAS Max % 100.0
Molecular Formula C4H3ClFN3
Packaging Glass bottle
CAS 155-10-2
Assay Percent Range 98%
Chemical Name or Material 4-Amino-2-chloro-5-fluoropyrimidine
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
Infrared Spectrum Authentic
MDL Number MFCD00057344
NMR Consistent with structure
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.