missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-2-chloro-3-iodopyridine, 95%

Manufacturer:  Alfa Aesar™ H33136MD

 View more versions of this product

Catalog No. AAH33136MD


 

Specifications

Specifications

4-Amino-2-chloro-3-iodopyridine
116°C to 117°C
250mg
95%
MFCD09763660
BYVMKCHHWASQFN-UHFFFAOYSA-N
2-chloro-3-iodopyridin-4-amine
24229208
Solid
909036-46-0
Orange-Pink to White
Light Sensitive
C5H4ClIN2
2-Chloro-3-iodo-4-pyridinamine
C1=CN=C(C(=C1N)I)Cl
254.455
254.46

Chemical Identifiers

909036-46-0
254.455
BYVMKCHHWASQFN-UHFFFAOYSA-N
24229208
C5H4ClIN2
MFCD09763660
2-Chloro-3-iodo-4-pyridinamine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

SDS Documents