Please login to your online account to display your discounted pricing

4-Amino-2,6-dihydroxypyrimidine 98%, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  103525000

 View more versions of this product

Catalog No. AC103525000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5% (HPLC)
Physical Form Crystalline Powder
Formula Weight 127.1
Quantity 500g
Color Cream to light brown
Beilstein 24, 469
EINECS Number 212-854-3
TSCA TSCA
RTECS Number YQ8750000
CAS Max % 100.0
Name Note 98%
Assay Percent Range 98%
Molecular Formula C4H5N3O2
Synonym 6-Amino-2, 4-pyrimidinediol
CAS Min % 97.5
Infrared Spectrum Authentic
Packaging Plastic bottle
Chemical Name or Material 4-Amino-2, 6-dihydroxypyrimidine
MDL Number MFCD00006071
CAS 873-83-6
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Amino-2, 6-dihydroxypyrimidine
Name Note 98%
CAS 873-83-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (HPLC)
Assay Percent Range 98%
Molecular Formula C4H5N3O2
Formula Weight 127.1
MDL Number MFCD00006071
Synonym 6-Amino-2, 4-pyrimidinediol
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents