Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-2,6-dichloropyrimidine, 98%

$137.26 - $549.97

Chemical Identifiers

Chemical Name or Material 4-Amino-2,6-dichloropyrimidine
CAS 10132-07-7
Molecular Formula C4H3Cl2N3
Formula Weight 164
MDL Number MFCD00038015
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1729406

  SDS
alfa aesar
A1729406
5g Each for $137.26

AAA1729414

  SDS
alfa aesar™
A1729414
25g Each for $549.97
Description & Specifications

Specifications

Formula Weight 164
Quantity 5g
Chemical Name or Material 4-Amino-2,6-dichloropyrimidine
TSCA TSCA
Melting Point 272° to 274°C
Assay 98%
Molecular Formula C4H3Cl2N3
MDL Number MFCD00038015
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 10132-07-7
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
It is used in the Suzuki coupling of 5-chloro-2-methoxyphenyl boronic acid with 4-amino-2,6-dichloropyrimidine yielded aminochloropyrimidine.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.