missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-2,6-dichloropyrimidine, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC443190050

 View more versions of this product

Catalog No. AC443190050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Amino-2, 6-dichloropyrimidine
Glass bottle
262.6°C to 263.2°C
10132-07-7
100.0
C4H3Cl2N3
UPVBKNZVOJNQKE-UHFFFAOYSA-N
2,6-dichloropyrimidin-4-amine
82387
Powder or Crystalline Powder
Authentic
Brown to White
5g
96.0
96% min. (HPLC)
2,4-dichloro-6-amino pyrimidine, 2,4-dichloro-6-aminopyrimidine, 2,6-dichloro-4-aminopyrimidine, 2,6-dichloro-pyrimidin-4-ylamine, 2,6-dichloropyrimidine-4-ylamine, 2,6-dichlorpyrimidin-4-amin, 4-amino-2,6-dichloropyrimidine, 4-pyrimidinamine, 2,6-dichloro, 6-amino-2,4-dichloropyrimidine, pyrimidine, 4-amino-2,6-dichloro
C1=C(N=C(N=C1Cl)Cl)N
163.99
163.99
97%

Chemical Identifiers

10132-07-7
163.99
2,4-dichloro-6-amino pyrimidine, 2,4-dichloro-6-aminopyrimidine, 2,6-dichloro-4-aminopyrimidine, 2,6-dichloro-pyrimidin-4-ylamine, 2,6-dichloropyrimidine-4-ylamine, 2,6-dichlorpyrimidin-4-amin, 4-amino-2,6-dichloropyrimidine, 4-pyrimidinamine, 2,6-dichloro, 6-amino-2,4-dichloropyrimidine, pyrimidine, 4-amino-2,6-dichloro
2,6-dichloropyrimidin-4-amine
C4H3Cl2N3
UPVBKNZVOJNQKE-UHFFFAOYSA-N
82387
C1=C(N=C(N=C1Cl)Cl)N
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning

EINECSNumber : 233-369-3

SDS Documents