missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-2,6-dichloropyridine, 98+%

Manufacturer:  Alfa Aesar™ A1503003

 View more versions of this product

Catalog No. AAA1503003



Specifications

Specifications

4-Amino-2,6-dichloropyridine
172°C to 176°C
2587-02-2
MFCD00052832
4-amino-2,6-dichloropyridine, 2,6-dichloro-4-aminopyridine, 2,6-dichloro-pyridin-4-ylamine, 4-pyridinamine, 2,6-dichloro, 2,6-dichloro-4-pyridylamine, 2,6-dichloropyridin-4-ylamine, 2,6-dichloro-4-amino pyridine, 2,6-dichloro-4-pyridinamine, pubchem5530, acmc-1cot2
C1=C(C=C(N=C1Cl)Cl)N
164.01
163.01
Unpleasant
1g
C5H5Cl2N2
114351
WAEZOSSWRXDWAX-UHFFFAOYSA-O
2,6-dichloropyridin-4-amine
137652
≥98%

Chemical Identifiers

2587-02-2
164.01
WAEZOSSWRXDWAX-UHFFFAOYSA-O
137652
C1=C(C=C(N=C1Cl)Cl)N
C5H5Cl2N2
MFCD00052832
4-amino-2,6-dichloropyridine, 2,6-dichloro-4-aminopyridine, 2,6-dichloro-pyridin-4-ylamine, 4-pyridinamine, 2,6-dichloro, 2,6-dichloro-4-pyridylamine, 2,6-dichloropyridin-4-ylamine, 2,6-dichloro-4-amino pyridine, 2,6-dichloro-4-pyridinamine, pubchem5530, acmc-1cot2
2,6-dichloropyridin-4-amine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Documents