missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-2,6-dichlorophenol 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 103481000

 View more versions of this product

Catalog No. AC103481000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
Powder
100g
164° to 168°C
Glass bottle
MFCD00007875
H2NC6H2(Cl)2OH
5930-28-9
4-Amino-2, 6-dichlorophenol
Beige or slightly brown to pale reddish
178.02
13, 512
100.0
C6H5Cl2NO
98%
98%
97.5
Authentic

Chemical Identifiers

5930-28-9
100.0
C6H5Cl2NO
178.02
4-Amino-2, 6-dichlorophenol, 98%
97.5
98%
H2NC6H2(Cl)2OH
MFCD00007875
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 227-671-4

TSCA : TSCA