missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-2,6-dichlorophenol 98%, ACROS Organics™

Manufacturer:  Acros Organics 103480250

 View more versions of this product

Catalog No. AC103480250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Amino-2, 6-dichlorophenol
Glass bottle
164°C to 168°C
98%
97.5
98%
H2NC6H2(Cl)2OH
13, 512
KGEXISHTCZHGFT-UHFFFAOYSA-N
4-amino-2,6-dichlorophenol
80037
Powder
Authentic
Beige or Brown to Red
25g
5930-28-9
100.0
C6H5Cl2NO
MFCD00007875
2,6-dichloro-4-amino phenol
C1=C(C=C(C(=C1Cl)O)Cl)N
178.012
178.02
≥97.5%

Chemical Identifiers

5930-28-9
178.012
KGEXISHTCZHGFT-UHFFFAOYSA-N
80037
C1=C(C=C(C(=C1Cl)O)Cl)N
C6H5Cl2NO
MFCD00007875
2,6-dichloro-4-amino phenol
4-amino-2,6-dichlorophenol
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 227-671-4

TSCA : TSCA

SDS Documents