Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-2,6-dibromophenol, 97%

$39.49 - $366.15

Chemical Identifiers

Chemical Name or Material 4-Amino-2,6-dibromophenol
CAS 609-21-2
Molecular Formula C6H5Br2NO
Formula Weight 266.93
MDL Number MFCD00007874
Beilstein 2803732
Synonym 4-Hydroxy-3,5-dibromoaniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1613406

  SDS
alfa aesar™
A1613406
5g Each for $39.49

AAA1613414

  SDS
alfa aesar™
A1613414
25g Each for $111.95

AAA1613422

  SDS
alfa aesar™
A1613422
100g Each for $366.15
Description & Specifications

Specifications

Formula Weight 266.93
Quantity 5g
TSCA TSCA
Assay 97%
CAS 609-21-2
Synonym 4-Hydroxy-3,5-dibromoaniline
Molecular Formula C6H5Br2NO
Health Hazard 1 Warning
MDL Number MFCD00007874
Chemical Name or Material 4-Amino-2,6-dibromophenol
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 195° to 196°C
Beilstein 2803732
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.