Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Amino-2',6'-dibromoacetanilide, 99%

$194.26 - $194.26

Chemical Identifiers

Chemical Name or Material 4'-Amino-2',6'-dibromoacetanilide
CAS 336624-08-9
Molecular Formula C8H8Br2N2O
Formula Weight 307.97g/mol
MDL Number MFCD02683078
Synonym 4-Acetamido-3,5-dibromoaniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2191506

 
alfa aesar™
B21915-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2191514

 
alfa aesar™
B21915-14
25g Each for $194.26
Description & Specifications

Specifications

Melting Point 246° to 249°C
Quantity 5g
Chemical Name or Material 4'-Amino-2',6'-dibromoacetanilide
CAS 336624-08-9
Assay 99%
Molecular Formula C8H8Br2N2O
Formula Weight 307.97g/mol
MDL Number MFCD02683078
Synonym 4-Acetamido-3,5-dibromoaniline
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.