missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-2,5-dimethylphenol, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC155330100

 View more versions of this product

Catalog No. AC155330100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Amino-2, 5-dimethylphenol
Glass bottle
242°C to 243°C
3096-71-7
100.0
C8H11NO
MFCD00007873
2,5-dimethy-4-aminophenol, 2,5-dimethyl-4-aminophenol, 2,5-xylenol, 4-amino, 4-amino-2,5-dimethyl-phenol, 4-amino-2,5-xylenol, 4-hydroxy-2,5-dimethylaniline, acmc-209hjo, ksc495q0r, phenol, 4-amino-2,5-dimethyl, unii-rf4r93709t
CC1=CC(=C(C=C1O)C)N
137.18
137.18
95%
Authentic
White to Beige
10g
94.0
94% min. (HPLC)
(CH3)2C6H2(OH)NH2
13, 634
JSWVCUXQICMATE-UHFFFAOYSA-N
4-amino-2,5-dimethylphenol
76544
Powder

Chemical Identifiers

3096-71-7
137.18
JSWVCUXQICMATE-UHFFFAOYSA-N
76544
CC1=CC(=C(C=C1O)C)N
C8H11NO
MFCD00007873
2,5-dimethy-4-aminophenol, 2,5-dimethyl-4-aminophenol, 2,5-xylenol, 4-amino, 4-amino-2,5-dimethyl-phenol, 4-amino-2,5-xylenol, 4-hydroxy-2,5-dimethylaniline, acmc-209hjo, ksc495q0r, phenol, 4-amino-2,5-dimethyl, unii-rf4r93709t
4-amino-2,5-dimethylphenol
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 221-449-

SDS Documents