missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-2,5-dimethylphenol, 95%, ACROS Organics™

Manufacturer:  Acros Organics AC155330050

 View more versions of this product

Catalog No. AC155330050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Amino-2, 5-dimethylphenol
94.0
Authentic
White through Beige
5g
C8H11NO
MFCD00007873
2, 5-Dimethyl-4-hydroxyaniline, 4-Amino-2, 5-xylenol
CC1=CC(=C(C=C1O)C)N
137.182
137.18
3096-71-7
100.0
Glass bottle
≥94%
95%
(CH3)2C6H2(OH)NH2
13, 634
JSWVCUXQICMATE-UHFFFAOYSA-N
4-amino-2,5-dimethylphenol
76544
Powder

Chemical Identifiers

3096-71-7
137.182
JSWVCUXQICMATE-UHFFFAOYSA-N
76544
C8H11NO
MFCD00007873
2, 5-Dimethyl-4-hydroxyaniline, 4-Amino-2, 5-xylenol
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents