missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-2,5-dimethylphenol, 95%, ACROS Organics™

Chemical Identifiers

CAS 3096-71-7
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00007873
InChI Key JSWVCUXQICMATE-UHFFFAOYSA-N
Synonym 2,5-dimethy-4-aminophenol
PubChem CID 76544
IUPAC Name 4-amino-2,5-dimethylphenol
SMILES CC1=CC(=C(C=C1O)C)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC155330050 SDS View Documents Thermo Fisher Scientific
AC155330050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC155330100 SDS View Documents Thermo Fisher Scientific
AC155330100
10g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

3096-71-7
137.18
JSWVCUXQICMATE-UHFFFAOYSA-N
76544
CC1=CC(=C(C=C1O)C)N
C8H11NO
MFCD00007873
2,5-dimethy-4-aminophenol
4-amino-2,5-dimethylphenol

Specifications

Authentic
White to Beige
5g
94.0
94% min. (HPLC)
(CH3)2C6H2(OH)NH2
13, 634
JSWVCUXQICMATE-UHFFFAOYSA-N
4-amino-2,5-dimethylphenol
76544
Powder
4-Amino-2, 5-dimethylphenol
Glass bottle
242°C to 243°C
3096-71-7
100.0
C8H11NO
MFCD00007873
2,5-dimethy-4-aminophenol
CC1=CC(=C(C=C1O)C)N
137.18
137.18
95%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 221-449-