Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-2,5-dimethoxy-N-phenylbenzenesulfonamide, 98%

Chemical Identifiers

Chemical Name or Material 4-Amino-2,5-dimethoxy-N-phenylbenzenesulfonamide
CAS 52298-44-9
Formula Weight 308.35
MDL Number MFCD00025753
Beilstein 13660911
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6639514

 
alfa aesar™
H6639514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6639522

 
alfa aesar™
H6639522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 308.35
Melting Point 184° to 188°C
Chemical Name or Material 4-Amino-2,5-dimethoxy-N-phenylbenzenesulfonamide
Physical Form Solid
MDL Number MFCD00025753
CAS 52298-44-9
Assay 98%
Beilstein 13660911
TSCA TSCA
Quantity 25g
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C