Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-2,3,5-tribromopyridine, 95%

$203.36 - $245.14

Chemical Identifiers

Chemical Name or Material 4-Amino-2,3,5-tribromopyridine
CAS 861024-31-9
Molecular Formula C5H3Br3N2
Formula Weight 330.80g/mol
MDL Number MFCD09037458
Synonym 2,3,5-Tribromo-4-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2769203

 
alfa aesar™
H27692-03
1g Each for $245.14

AAH27692MD

 
alfa aesar™
H27692-MD
250mg Each for $203.36
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material 4-Amino-2,3,5-tribromopyridine
CAS 861024-31-9
Assay 95%
Molecular Formula C5H3Br3N2
Formula Weight 330.80g/mol
MDL Number MFCD09037458
Synonym 2,3,5-Tribromo-4-pyridinamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.