Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-2,3,5,6-tetrafluoropyridine, 98%

$64.80 - $265.19

Chemical Identifiers

Chemical Name or Material 4-Amino-2,3,5,6-tetrafluoropyridine
CAS 1682-20-8
Molecular Formula C5H2F4N2
Formula Weight 166.08
MDL Number MFCD00010855
Beilstein 475870
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2381503

  SDS
alfa aesar™
B2381503
1g Each for $64.80

AAB2381506

  SDS
alfa aesar™
B2381506
5g Each for $265.19
Description & Specifications

Specifications

Formula Weight 166.08
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Danger
Molecular Formula C5H2F4N2
Melting Point 84° to 87°C
CAS 1682-20-8
Quantity 1g
MDL Number MFCD00010855
Assay 98%
Chemical Name or Material 4-Amino-2,3,5,6-tetrafluoropyridine
Beilstein 475870
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.