Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-1H-pyrazolo[3,4-d]pyrimidine, 98%

Chemical Identifiers

Chemical Name or Material 4-Amino-1H-pyrazolo[3,4-d]pyrimidine
CAS 2380-63-4
Molecular Formula C5H5N5
Formula Weight 135.13
MDL Number MFCD00005688
UN Number UN2811
Beilstein 5824
Synonym 1H-Pyrazolo[3,4-d]pyrimidin-4-amine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0351903

  SDS
alfa aesar™
L0351903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL03519MD

  SDS
alfa aesar™
L03519MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 135.13
Chemical Name or Material 4-Amino-1H-pyrazolo[3,4-d]pyrimidine
TSCA TSCA
MDL Number MFCD00005688
CAS 2380-63-4
UN Number UN2811
Quantity 1g
Synonym 1H-Pyrazolo[3,4-d]pyrimidin-4-amine
Assay 98%
Molecular Formula C5H5N5
Melting Point >310°C
Beilstein 5824
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number UR0717000
Odor Odorless

Applications
4-Amino-1H-pyrazolo[3,4-d]pyrimidine decreases serum cholesterol markedly in rats.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.