missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-1H-isoindole-1,3(2H)-dione, 97%, ACROS Organics™

Chemical Identifiers

CAS 2518-24-3
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C8H6N2O2
Formula Weight 162.15
Synonym 4-Amino-1H-isoindole-1, 3(2H)-dione
Chemical Name or Material 3-Aminophthalimide
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC401000010 SDS Thermo Fisher Scientific
401000010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

2518-24-3
100.0
C8H6N2O2
4-Amino-1H-isoindole-1, 3(2H)-dione
≥98.5%
98.5
99%
162.15
3-Aminophthalimide

Specifications

162.15
1g
100.0
4-Amino-1H-isoindole-1, 3(2H)-dione
2518-24-3
Authentic
3-Aminophthalimide
Fine Powder
Yellow
Glass bottle
99%
98.5
C8H6N2O2
≥98.5%