missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-1H-isoindole-1,3(2H)-dione, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 401000010

 View more versions of this product

Catalog No. AC401000010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5%
Fine Powder
Yellow
Glass bottle
99%
2518-24-3
Authentic
162.15
1g
100.0
4-Amino-1H-isoindole-1, 3(2H)-dione
3-Aminophthalimide
98.5
C8H6N2O2

Chemical Identifiers

2518-24-3
100.0
C8H6N2O2
4-Amino-1H-isoindole-1, 3(2H)-dione
≥98.5%
98.5
99%
162.15
3-Aminophthalimide