Please login to your online account to display your discounted pricing

4-amino-1-phenylpyrazole[3,4-d]pyrimidine, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Amino-1-phenylpyrazole[3, 4-d]pyrimidine
CAS 5334-30-5
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C11H9N5
Formula Weight 211.22
MDL Number MFCD00022901
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC374140050

  SDS
thermo fisher scientific
AC374140050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 211.22
Quantity 5g
Color Beige
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 4-Amino-1-phenylpyrazole[3, 4-d]pyrimidine
Health Hazard 1 Warning
MDL Number MFCD00022901
Assay Percent Range 95%
CAS 5334-30-5
Infrared Spectrum Authentic
Molecular Formula C11H9N5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.