Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-1-phenyl-1H-pyrazolo[3,4-d]pyrimidine, tech. 85%

$103.66 - $385.73

Chemical Identifiers

Chemical Name or Material 4-Amino-1-phenyl-1H-pyrazolo[3,4-d]pyrimidine
CAS 5334-30-5
Purity Grade Technical Grade
Molecular Formula C11H9N5
Formula Weight 211.23
MDL Number MFCD00022901
Beilstein 198460
Synonym 1-Phenyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0712506

  SDS
alfa aesar™
L0712506
5g Each for $385.73

AAL0712503

  SDS
alfa aesar™
L0712503
1g Each for $103.66
Description & Specifications

Specifications

Chemical Name or Material 4-Amino-1-phenyl-1H-pyrazolo[3,4-d]pyrimidine
CAS 5334-30-5
Melting Point 214° to 217°C
Purity Grade Technical Grade
Assay 85%
Quantity 5g
Molecular Formula C11H9N5
Formula Weight 211.23
MDL Number MFCD00022901
Beilstein 198460
Synonym 1-Phenyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine