missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Amino-1-naphthol Hydrochloride (Tech.), 90%, ACROS Organics™

Manufacturer:  Acros Organics 158510250

 View more versions of this product

Catalog No. AC158510250



Specifications

Specifications

4-Amino-1-naphthol hydrochloride
Glass bottle
273.0°C
tech., 90%
88.0
90%
H2NC10H6OH·HCl
13, 667
4-amino-1-naphthol hydrochloride, 4-aminonaphthalen-1-ol hydrochloride, 1-amino-4-naphthol hydrochloride, 1-naphthalenol, 4-amino-, hydrochloride, 4-amino-1-naphtholhydrochloride, 4-aminonaphthol, chloride, pcmd-cc-pab-253, 1-naphthalenol, 4-amino-, hydrochloride 1:1, acmc-1aogt, 4-amino-1-naphthol . hcl
C1=CC=C2C(=C1)C(=CC=C2O)N.Cl
195.646
195.65
90%
Authentic
Gray to Pink
25g
5959-56-8
100.0
C10H10ClNO
MFCD00012562
10, 457
FDBQTRARWCKEJY-UHFFFAOYSA-N
4-aminonaphthalen-1-ol;hydrochloride
2723858
Powder
Technical

Chemical Identifiers

5959-56-8
195.646
FDBQTRARWCKEJY-UHFFFAOYSA-N
2723858
C1=CC=C2C(=C1)C(=CC=C2O)N.Cl
C10H10ClNO
MFCD00012562
4-amino-1-naphthol hydrochloride, 4-aminonaphthalen-1-ol hydrochloride, 1-amino-4-naphthol hydrochloride, 1-naphthalenol, 4-amino-, hydrochloride, 4-amino-1-naphtholhydrochloride, 4-aminonaphthol, chloride, pcmd-cc-pab-253, 1-naphthalenol, 4-amino-, hydrochloride 1:1, acmc-1aogt, 4-amino-1-naphthol . hcl
4-aminonaphthalen-1-ol;hydrochloride
Safety and Handling

Safety and Handling

EINECSNumber : 227-727-8

RTECSNumber : QL3342000

TSCA : TSCA

SDS Documents