missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
4-Amino-1-isopropylpiperidine, TCI America™
Chemical Identifiers
CAS |
127285-08-9 |
Molecular Formula |
C8H18N2 |
Molecular Weight (g/mol) |
142.246 |
MDL Number |
MFCD03411606 |
InChI Key |
ZRQQXFMGYSOKDF-UHFFFAOYSA-N |
PubChem CID |
3163333 |
IUPAC Name |
1-propan-2-ylpiperidin-4-amine |
SMILES |
CC(C)N1CCC(CC1)N |
View More Specs
Products
2
Specifications
Chemical Identifiers
|
127285-08-9 |
|
142.246 |
|
ZRQQXFMGYSOKDF-UHFFFAOYSA-N |
|
1-propan-2-ylpiperidin-4-amine |
|
C8H18N2 |
|
MFCD03411606 |
|
3163333 |
|
CC(C)N1CCC(CC1)N |
Specifications
|
Colorless |
|
127285-08-9 |
|
MFCD03411606 |
|
ZRQQXFMGYSOKDF-UHFFFAOYSA-N |
|
1-propan-2-ylpiperidin-4-amine |
|
3163333 |
|
Clear Liquid at 20°C |
|
4-Amino-1-isopropylpiperidine |
|
5g |
|
C8H18N2 |
|
2920 |
|
CC(C)N1CCC(CC1)N |
|
142.246 |
|
142.25 |
|
≥98.0% (GC,T) |
Safety and Handling