Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-1-isobutyrylpiperidine, 95%

$321.72 - $321.72

Chemical Identifiers

Chemical Name or Material 4-Amino-1-isobutyrylpiperidine
CAS 172478-05-6
Molecular Formula C9H18N2O
Formula Weight 170.26
MDL Number MFCD08061057
Synonym 1-Isobutyryl-4-piperidinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5091603

  SDS
alfa aesar™
H5091603
1g Each for $321.72
Description & Specifications

Specifications

Formula Weight 170.26
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 4-Amino-1-isobutyrylpiperidine
Assay 95%
Molecular Formula C9H18N2O
MDL Number MFCD08061057
CAS 172478-05-6
Synonym 1-Isobutyryl-4-piperidinamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.