Please login to your online account to display your discounted pricing

4-Amino-1-benzylpiperidine dihydrochloride hydrate, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Amino-1-benzylpiperidine dihydrochloride hydrate
Name Note 95%
CAS 1205-72-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C12 H18 N2.2 H Cl.x H2 O
Formula Weight 263.2
MDL Number MFCD00167361
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC135172500

  SDS
thermo fisher scientific
135172500
Glass bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Fine Crystalline Powder
Formula Weight 263.2
Quantity 250g
Color White to very slightly yellow
EINECS Number 214-886-3
CAS Min % 94.0
CAS Max % 100.0
Molecular Formula C12 H18 N2.2 H Cl.x H2 O
Chemical Name or Material 4-Amino-1-benzylpiperidine dihydrochloride hydrate
Packaging Glass bottle
Name Note 95%
Assay Percent Range 95%
MDL Number MFCD00167361
CAS 1205-72-7
Infrared Spectrum Authentic