Please login to your online account to display your discounted pricing

4-Amino-1-benzylpiperidine dihydrochloride hydrate, 95%, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  135172500

 View more versions of this product

Catalog No. AC135172500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Fine Crystalline Powder
Formula Weight 263.2
Quantity 250g
Color White to very slightly yellow
EINECS Number 214-886-3
CAS Min % 94.0
CAS Max % 100.0
Molecular Formula C12 H18 N2.2 H Cl.x H2 O
MDL Number MFCD00167361
Assay Percent Range 95%
Name Note 95%
Chemical Name or Material 4-Amino-1-benzylpiperidine dihydrochloride hydrate
CAS 1205-72-7
Packaging Glass bottle
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 4-Amino-1-benzylpiperidine dihydrochloride hydrate
Name Note 95%
CAS 1205-72-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C12 H18 N2.2 H Cl.x H2 O
Formula Weight 263.2
MDL Number MFCD00167361