Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-1-benzylpiperidine, 98%

$51.08 - $411.35

Chemical Identifiers

Chemical Name or Material 4-Amino-1-benzylpiperidine
CAS 50541-93-0
Molecular Formula C12H18N2
Formula Weight 190.29
MDL Number MFCD00006504
Beilstein 146038
Synonym 1-Benzyl-4-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1014906

  SDS
alfa aesar™
A1014906
5g Each for $51.08

AAA1014914

  SDS
alfa aesar™
A1014914
25g Each for $152.19

AAA1014922

  SDS
alfa aesar™
A1014922
100g Each for $411.35
Description & Specifications

Specifications

Formula Weight 190.29
Chemical Name or Material 4-Amino-1-benzylpiperidine
Assay 98%
Molecular Formula C12H18N2
Boiling Point 279° to 281°C
Quantity 5g
Flash Point >110°C (230°F)
MDL Number MFCD00006504
CAS 50541-93-0
Beilstein 146038
Synonym 1-Benzyl-4-pyridinamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.