missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-1-benzylpiperidine, 98%

Manufacturer:  Alfa Aesar™ A1014914

 View more versions of this product

Catalog No. AAA1014914



Specifications

Specifications

4-Amino-1-benzylpiperidine
279°C to 281°C
50541-93-0
MFCD00006504
4-amino-1-benzylpiperidine, 1-benzyl-4-aminopiperidine, 1-benzyl-piperidin-4-ylamine, n-benzyl-4-amino piperidine, 1-benzyl-4-piperidylamine, 4-piperidinamine, 1-phenylmethyl, 4-amino-1-n-benzylpiperidine, 4-amino-n-benzylpiperidine, 1-phenylmethyl-4-piperidinamine, n-benzyl-4-aminopiperidine
C1CN(CCC1N)CC2=CC=CC=C2
190.29
190.29
>110°C (230°F)
25g
C12H18N2
146038
YUBDLZGUSSWQSS-UHFFFAOYSA-N
1-benzylpiperidin-4-amine
415852
98%

Chemical Identifiers

50541-93-0
190.29
YUBDLZGUSSWQSS-UHFFFAOYSA-N
415852
C1CN(CCC1N)CC2=CC=CC=C2
C12H18N2
MFCD00006504
4-amino-1-benzylpiperidine, 1-benzyl-4-aminopiperidine, 1-benzyl-piperidin-4-ylamine, n-benzyl-4-amino piperidine, 1-benzyl-4-piperidylamine, 4-piperidinamine, 1-phenylmethyl, 4-amino-1-n-benzylpiperidine, 4-amino-n-benzylpiperidine, 1-phenylmethyl-4-piperidinamine, n-benzyl-4-aminopiperidine
1-benzylpiperidin-4-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

SDS Documents