Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-1-(3-methyl-2-pyridyl)piperidine, 99%

$133.80 - $465.59

Chemical Identifiers

Chemical Name or Material 4-Amino-1-(3-methyl-2-pyridyl)piperidine
CAS 902837-09-6
Molecular Formula C11H17N3
Formula Weight 191.28
MDL Number MFCD08060981
Synonym 1-(3-Methylpyridin-2-yl)piperidin-4-amine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5077903

  SDS
alfa aesar™
H5077903
1g Each for $465.59

AAH50779MD

  SDS
alfa aesar™
H50779MD
250mg Each for $133.80
Description & Specifications

Specifications

Formula Weight 191.28
Molecular Formula C11H17N3
Assay 99%
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 4-Amino-1-(3-methyl-2-pyridyl)piperidine
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Danger
Quantity 1g
Synonym 1-(3-Methylpyridin-2-yl)piperidin-4-amine
MDL Number MFCD08060981
CAS 902837-09-6
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.