Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-1-(3-methoxy-2-pyridyl)piperidine hydrochloride

$110.00 - $110.00

Chemical Identifiers

Chemical Name or Material 4-Amino-1-(3-methoxy-2-pyridyl)piperidine hydrochloride
Molecular Formula C11H17N3O·HCl
Formula Weight 243.73
MDL Number MFCD11869912
Synonym 1-(3-Methoxy-2-pyridyl)-4-piperidinamine hydrochloride; 2-(4-Amino-1-piperidinyl)-3-methoxypyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5117603

  SDS
alfa aesar™
H5117603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH51176MD

  SDS
alfa aesar™
H51176MD
250mg Each for $110.00
Description & Specifications

Specifications

Formula Weight 243.73
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 4-Amino-1-(3-methoxy-2-pyridyl)piperidine hydrochloride
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C11H17N3O·HCl
MDL Number MFCD11869912
Quantity 1g
Health Hazard 1 Danger
Synonym 1-(3-Methoxy-2-pyridyl)-4-piperidinamine hydrochloride; 2-(4-Amino-1-piperidinyl)-3-methoxypyridine
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.