Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-1-(3-chloro-2-pyridyl)piperidine hydrochloride, 97%

$160.08 - $547.99

Chemical Identifiers

Chemical Name or Material 4-Amino-1-(3-chloro-2-pyridyl)piperidine hydrochloride
CAS 77145-35-8
Molecular Formula C10H14ClN3·HCl
Formula Weight 248.15
MDL Number MFCD16251586
Synonym 1-(3-Chloro-2-pyridyl)piperidin-4-amine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5192703

  SDS
alfa aesar™
H5192703
1g Each for $547.99

AAH51927MD

  SDS
alfa aesar™
H51927MD
250mg Each for $160.08
Description & Specifications

Specifications

Formula Weight 248.15
Chemical Name or Material 4-Amino-1-(3-chloro-2-pyridyl)piperidine hydrochloride
Melting Point 265° to 267°C
Quantity 1g
Assay 97%
Molecular Formula C10H14ClN3·HCl
MDL Number MFCD16251586
CAS 77145-35-8
Synonym 1-(3-Chloro-2-pyridyl)piperidin-4-amine hydrochloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.