Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Amino-1,2,4-triazole, 99%

$39.70 - $273.99

Chemical Identifiers

Chemical Name or Material 4-Amino-1,2,4-triazole
CAS 584-13-4
Molecular Formula C2H4N4
Formula Weight 84.08
MDL Number MFCD00003099
Beilstein 107563
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1771606

  SDS
alfa aesar™
A1771606
5g Each for $39.70

AAA1771614

  SDS
alfa aesar™
A1771614
25g Each for $108.07

AAA1771622

  SDS
alfa aesar™
A1771622
100g Each for $273.99
Description & Specifications

Specifications

Formula Weight 84.08
Chemical Name or Material 4-Amino-1,2,4-triazole
Assay 99%
Molecular Formula C2H4N4
Melting Point 83° to 88°C
MDL Number MFCD00003099
CAS 584-13-4
Quantity 5g
Beilstein 107563
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number XZ3850200

Applications
4-Amino-1,2,4-triazole is used as an intermediate for the preparation of antifungal agent like fluconazole.

Solubility
Soluble in water and methanol.

Notes
Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.