missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(alpha-Methylbenzyl)resorcinol, TCI America™

$76.39 - $252.09

Chemical Identifiers

CAS 85-27-8
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.264
MDL Number MFCD21648466
InChI Key PQSXNIMHIHYFEE-UHFFFAOYSA-N
Synonym 4-(1-Phenylethyl)benzene-1,3-diol, 4-(1-Phenylethyl)resorcinol
PubChem CID 11171903
IUPAC Name 4-(1-phenylethyl)benzene-1,3-diol
SMILES CC(C1=CC=CC=C1)C2=C(C=C(C=C2)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M27545G View Documents TCI America
M27545G
5g Each for $252.09
 
M27541G View Documents TCI America
M27541G
1g Each for $76.39
 
Specifications

Chemical Identifiers

85-27-8
214.264
PQSXNIMHIHYFEE-UHFFFAOYSA-N
11171903
CC(C1=CC=CC=C1)C2=C(C=C(C=C2)O)O
C14H14O2
MFCD21648466
4-(1-Phenylethyl)benzene-1,3-diol, 4-(1-Phenylethyl)resorcinol
4-(1-phenylethyl)benzene-1,3-diol

Specifications

244°C
81°C
85-27-8
MFCD21648466
PQSXNIMHIHYFEE-UHFFFAOYSA-N
4-(1-phenylethyl)benzene-1,3-diol
11171903
Crystal-Powder at 20°C
4-(alpha-Methylbenzyl)resorcinol
Undesignated
5g
C14H14O2
4-(1-Phenylethyl)benzene-1,3-diol, 4-(1-Phenylethyl)resorcinol
CC(C1=CC=CC=C1)C2=C(C=C(C=C2)O)O
214.264
214.26
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No