Please login to your online account to display your discounted pricing

4-Allyloxybenzaldehyde 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC391810250

 View more versions of this product

Catalog No. AC391810250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Viscous Liquid
Formula Weight 162.19
Quantity 25g
Color Clear slightly yellow
CAS Max % 100.0
Chemical Name or Material 4-Allyloxybenzaldehyde
Packaging Glass bottle
Refractive Index 1.5660 to 1.5680 (20°C, 589nm)
Assay Percent Range 98%
Molecular Formula C10H10O2
Linear Formula CH2CHCH2OC6H4CHO
MDL Number MFCD00014133
CAS 40663-68-1
CAS Min % 97.5
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

Specifications

Chemical Name or Material 4-Allyloxybenzaldehyde
CAS 40663-68-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H10O2
Linear Formula CH2CHCH2OC6H4CHO
Formula Weight 162.19
MDL Number MFCD00014133
Accessories

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents