missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Allyloxybenzaldehyde 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC391810050

 View more versions of this product

Catalog No. AC391810050


 

Specifications Description & Specifications

Specifications

4-Allyloxybenzaldehyde
97.5
Authentic
1.5660 to 1.5680 (20°C, 589nm)
≥97.5%
C10H10O2
162.19
Clear slightly yellow
40663-68-1
100.0
Glass bottle
Viscous Liquid
98%
CH2CHCH2OC6H4CHO
MFCD00014133
5g

Chemical Identifiers

40663-68-1
100.0
C10H10O2
162.19
4-Allyloxybenzaldehyde
97.5
98%
CH2CHCH2OC6H4CHO
MFCD00014133
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.