Please login to your online account to display your discounted pricing

4-Allyloxybenzaldehyde 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Allyloxybenzaldehyde
CAS 40663-68-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H10O2
Linear Formula CH2CHCH2OC6H4CHO
Formula Weight 162.19
MDL Number MFCD00014133
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC391810050

  SDS
thermo fisher scientific
AC391810050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC391810250

  SDS
thermo fisher scientific
AC391810250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 162.19
Physical Form Viscous Liquid
Quantity 5g
Color Clear slightly yellow
CAS Max % 100.0
Linear Formula CH2CHCH2OC6H4CHO
CAS 40663-68-1
Assay Percent Range 98%
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00014133
CAS Min % 97.5
Infrared Spectrum Authentic
Chemical Name or Material 4-Allyloxybenzaldehyde
Packaging Glass bottle
Refractive Index 1.5660 to 1.5680 (20°C, 589nm)
Molecular Formula C10H10O2
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Safety and Handling

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.