missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Allylanisole 98%, ACROS Organics™

Manufacturer:  Acros Organics 102880050

 View more versions of this product

Catalog No. AC102880050



Specifications

Specifications

4-Allylanisole
Glass bottle
Colorless
98%
97.5
98%
H2C=CHCH2C6H4OCH3
06, 571
1-allyl-4-methoxybenzene
COC1=CC=C(C=C1)CC=C
148.205
CHEBI:4867
Liquid
Authentic
1.5200 to 1.5240 (20°C, 589nm)
5g
140-67-0
100.0
C10H12O
MFCD00008653
15, 3760
ZFMSMUAANRJZFM-UHFFFAOYSA-N
1-methoxy-4-prop-2-enylbenzene
8815
148.2
≥97.5%

Chemical Identifiers

140-67-0
148.205
ZFMSMUAANRJZFM-UHFFFAOYSA-N
8815
1-methoxy-4-prop-2-enylbenzene
C10H12O
MFCD00008653
1-allyl-4-methoxybenzene
CHEBI:4867
COC1=CC=C(C=C1)CC=C
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 205-427-8

RTECSNumber : BZ8225000

TSCA : TSCA

SDS Documents