missing translation for 'onlineSavingsMsg'
Learn More

4-Acetyltetrahydropyran, 97%

CAS: 137052-08-5 | C7H12O2 | 128.171 g/mol

Supplier:  Thermo Scientific Chemicals H6452203

 View more versions of this product

Catalog No. AAH6452203


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Acetyltetrahydropyran
Colorless to Yellow
90°C to 94°C (15 mmHg)
1.453
1 g
1-tetrahydro-2h-pyran-4-yl ethanone, 1-oxan-4-yl ethan-1-one, 1-oxan-4-yl ethanone, 4-acetyltetrahydropyran, 1-tetrahydro-2h-pyran-4-ylethanone, 4-acetyltetrahydro-4h-pyran, 4-acetyltetrahydro-2h-pyran, 1-tetrahydropyran-4-ylethanone, 1-tetrahydro-pyran-4-yl-ethanone, 1-tetrahydro-2h-pyran-4-yl ethan-1-one
CC(=O)C1CCOCC1
128.171
128.17
Liquid
137052-08-5
1.024 g/mL
C7H12O2
MFCD08704647
109587
VNMXIOWPBADSIC-UHFFFAOYSA-N
1-(oxan-4-yl)ethanone
9877365
97%

Chemical Identifiers

137052-08-5
128.171
VNMXIOWPBADSIC-UHFFFAOYSA-N
9877365
CC(=O)C1CCOCC1
C7H12O2
MFCD08704647
1-tetrahydro-2h-pyran-4-yl ethanone, 1-oxan-4-yl ethan-1-one, 1-oxan-4-yl ethanone, 4-acetyltetrahydropyran, 1-tetrahydro-2h-pyran-4-ylethanone, 4-acetyltetrahydro-4h-pyran, 4-acetyltetrahydro-2h-pyran, 1-tetrahydropyran-4-ylethanone, 1-tetrahydro-pyran-4-yl-ethanone, 1-tetrahydro-2h-pyran-4-yl ethan-1-one
1-(oxan-4-yl)ethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only