missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Acetylpyridine, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC102705000

 View more versions of this product

Catalog No. AC102705000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Acetylpyridine
104°C
Glass bottle
1.09
Brown to Yellow
500g
97.5
98%
(C5H4N)COCH3
21, 279
Solubility in water: insoluble
CC(=O)C1=CC=NC=C1
121.14
121.14
98%
1.0900g/mL
Authentic
1.5270 to 1.5310
212.0°C
13.0°C to 16.0°C
1122-54-9
100.0
C7H7NO
MFCD00006433
1-pyridin-4-yl ethan-1-one
WMQUKDQWMMOHSA-UHFFFAOYSA-N
1-pyridin-4-ylethanone
14282
Liquid

Chemical Identifiers

1122-54-9
121.14
WMQUKDQWMMOHSA-UHFFFAOYSA-N
14282
CC(=O)C1=CC=NC=C1
C7H7NO
MFCD00006433
1-pyridin-4-yl ethan-1-one
1-pyridin-4-ylethanone
Safety and Handling

Safety and Handling

GHS H StatementCauses serious eye irritation.May cause respiratory irritation.Causes skin irritation.

GHS P StatementIF ON SKIN: Wash with plenty of soap and water.If skin irritation occurs: Get medical advice/attention.If eye irritation persists: Get medical advice/attention.IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.Call a POISON CENTER or doctor/physician if you feel unwell.Wear protective gloves/protective clothing/eye protection/face protection.

;GHS P StatementIF ON SKIN: Wash with plenty of soap and water.If skin irritation occurs: Get medical advice/attention.If eye irritation persists: Get medical advice/attention.IF INHALED: Remove to fresh air and keep at rest in a position c

SDS Documents