missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetylpyridine, 98%

Manufacturer:  Alfa Aesar™ A1201536

 View more versions of this product

Catalog No. AAA1201536


 

Description

Description

Applications
4-acetylpyridine on reaction with - and 4-formylpyridine (H3FoPyS, H4FoPyS) yields semicarbazones. Reaction of acetylpyridine with thiophene-2-carboxaldehyde afforded the 2-chloro-6-ethoxy-4-β-(2-thienyl)acryloylpyridine, which was reacted with malononitrile in refluxing ethanol in the presence of piperidine as a catalyst to afford the cyanoaminopyrane derivative.

Solubility
Insoluble in water. Soluble in alcohol.

Notes
Store in cool place. Keep away from oxidizing agents.
Specifications

Specifications

4-Acetylpyridine
98%
MFCD00006433
Methyl 4-pyridyl ketone
CC(=O)C1=CC=NC=C1
121.139
121.14
210°C to 212°C
104°C (219°F)
1122-54-9
C7H7NO
107629
WMQUKDQWMMOHSA-UHFFFAOYSA-N
1-pyridin-4-ylethanone
14282
14°C to 16°C
500g

Chemical Identifiers

1122-54-9
121.139
WMQUKDQWMMOHSA-UHFFFAOYSA-N
14282
C7H7NO
MFCD00006433
Methyl 4-pyridyl ketone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : OB5426000

TSCA : TSCA

SDS Documents