Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Acetylpyridazine, 95%

Chemical Identifiers

Chemical Name or Material 4-Acetylpyridazine
CAS 50901-46-7
Formula Weight 122.13
MDL Number MFCD13193354
Beilstein 742138
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH66461MD

  SDS
alfa aesar™
H66461MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6646103

  SDS
alfa aesar™
H6646103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 122.13
Chemical Name or Material 4-Acetylpyridazine
Assay 95%
CAS 50901-46-7
Physical Form Solid
MDL Number MFCD13193354
Storage Note 1 -30° to -10°C
Beilstein 742138
Quantity 250mg
Safety and Handling

-30° to -10°C