Please login to your online account to display your discounted pricing

4-Acetylphenylboronic acid pinacol ester, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Acetylphenylboronic acid pinacol ester
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C14H19BO3
Formula Weight 246.12
Synonym 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC444430010

 
thermo fisher scientific
AC444430010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC444430050

 
thermo fisher scientific
AC444430050
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥96.0% (GC)
Formula Weight 246.12
Quantity 1g
Color White
CAS 171364-81-1
Infrared Spectrum Authentic
Assay Percent Range 97%
NMR 97 % min. consistent with structure (1H-NMR)
Molecular Formula C14H19BO3
Chemical Name or Material 4-Acetylphenylboronic acid pinacol ester
Synonym 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone