missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Acetylphenylboronic acid pinacol ester, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 444430010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Acetylphenylboronic acid pinacol ester | |
80°C (0.1 Torr) | |
97% | |
C14H19BO3 | |
1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone, 4-acetylphenylboronic acid pinacol ester, 4-acetylphenylboronic acid, pinacol ester, 2-4-acetylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethan-1-one, amtb682, 4-acetylphenyl boronic acid pinacol ester, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2yl phenyl-ethanone | |
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)C | |
246.12 | |
246.12 |
171364-81-1 | |
Authentic | |
Glass Bottle | |
1g | |
BATKIZWNRQGSKE-UHFFFAOYSA-N | |
1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone | |
2760596 | |
97% |
Chemical Identifiers
171364-81-1 | |
246.12 | |
1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone, 4-acetylphenylboronic acid pinacol ester, 4-acetylphenylboronic acid, pinacol ester, 2-4-acetylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethan-1-one, amtb682, 4-acetylphenyl boronic acid pinacol ester, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl ethanone, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2yl phenyl-ethanone | |
1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]ethanone |
C14H19BO3 | |
BATKIZWNRQGSKE-UHFFFAOYSA-N | |
2760596 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)C |
Safety and Handling
GHS Signal Word: Warning
RUO â Research Use Only